missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trimethoxybenzoic acid, 99%

Product Code. p-7024519
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11479576

Brand: Thermo Scientific Alfa Aesar A19324.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 490-64-2 | C10H11O5 | 211.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 490-64-2
Molecular Formula C10H11O5
Molecular Weight (g/mol) 211.19
MDL Number MFCD00002435
InChI Key KVZUCOGWKYOPID-UHFFFAOYSA-M
Synonym asaronic acid, benzoic acid, 2,4,5-trimethoxy, asarylic acid, calamonic acid, acmc-209ket, 2,4,5-trimethoxybenzoicacid, ksc489o7d, 2,4,5-trimethoxy-benzoic acid, 2,4,5-trimethoxybenzoic acid, alpha-hydroxyphenylacetic acid; amygdalic acid; alpha-hydroxyphenylacetic acid; alpha-hydroxy-benzeneacetic acid
PubChem CID 10276
SMILES COC1=CC(OC)=C(C=C1OC)C([O-])=O

Specifications

Melting Point 143°C to 145°C
Quantity 10 g
IUPAC Name 2,4,5-trimethoxybenzoate
Formula Weight 212.2
Percent Purity 99%
Chemical Name or Material 2,4,5-Trimethoxybenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.