missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenoxy)-2-methylpropionic acid 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30311998
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30311998 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311998 Supplier Dr. Ehrenstorfer Supplier No. DREA11484000AL100
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(4-Chlorophenoxy)-2-methylpropionic acid (Clofibric acid)
CAS 882-09-7
Analyte or Component Names 2-(4-Chlorophenoxy)-2-methylpropionic acid (Clofibric acid)
Shipping Condition +20°C
Molecular Formula C10 H11 Cl O3
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Propanoic acid, 2-(4-chlorophenoxy)-2-methyl-, Propionic acid, 2-(p-chlorophenoxy)-2-methyl- (6CI,7CI,8CI), 2-(4-Chlorophenoxy)-2-methylpropanoic acid, (p-Chlorophenoxy)isobutyric acid, 2-(4-Chlorophenoxy)-2-methylpropanic acid, 2-(4-Chlorophenoxy)-2-methylpropionic acid, 2-(p-Chlorophenoxy)-2-methylpropanoic acid, 2-(p-Chlorophenoxy)-2-methylpropionic acid, 2-(p-Chlorophenoxy)isobutyric acid, 2-[(4-Chlorophenyl)oxy]-2-methylpropionic acid, 4-Chlorophenoxyisobutyric acid, Arteriohom, CPIB, CPIBA, Chlorfibrinic acid, Chlorofibrinic acid, Chlorophibrinic acid, Clofibrate free acid, Clofibric acid, Clofibrilic acid, Clofibrin, Clofibrinic acid, NSC 1149, PCIB, PCPIB, Regadrin, Regulipid, α-(4-Chlorophenoxy)-α-methylpropionic acid, α-(4-Chlorophenoxy)isobutyric acid, α-(p-Chlorophenoxy)isobutyric acid
InChI Formula InChI=1S/C10H11ClO3/c1-10(2,9(12)13)14-8-5-3-7(11)4-6-8/h3-6H,1-2H3,(H,12,13)
SMILES CC(C)(Oc1ccc(Cl)cc1)C(=O)O
IUPAC Name 2-(4-chlorophenoxy)-2-methylpropanoic acid
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 214.645
Quantity 1 mL
Formula Weight 214.04 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.