missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Morpholinyl)ethanol, Mikromol™

Product Code. 30051519
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051519 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051519 Supplier Mikromol Supplier No. MM1001.110100
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-(4-Morpholinyl)ethanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Morpholineethanol
CAS 622-40-2
Analyte or Component Names 4-Morpholineethanol
Shipping Condition Room Temperature
Molecular Formula C6 H13 N O2
Certifications/Compliance ISO 9001
Synonym 4-Morpholineethanol, 2-(4-Morpholino)ethyl alcohol, 2-(4-Morpholinyl)-1-ethanol, 2-(4-Morpholinyl)ethanol, 2-Morpholinoethanol, 4-(2-Hydroxyethyl)morpholine, N-(2-Hydroxyethyl)morpholine, N-(β-Hydroxyethyl)morpholine, NSC 1946, β-Morpholinoethanol
InChI Formula InChI=1S/C6H13NO2/c8-4-1-7-2-5-9-6-3-7/h8H,1-6H2
SMILES OCCN1CCOCC1
IUPAC Name 2-morpholin-4-ylethanol
Molecular Weight (g/mol) 131.17
Quantity 100 mg
Formula Weight 131.0946 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.