missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis-(trifluoromethyl)aniline, Mikromol™

Product Code. 30051227
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051227 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051227 Supplier Mikromol Supplier No. MM1424.01
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,5-Bis-(trifluoromethyl)aniline
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,5-Bis-(trifluoromethyl)aniline
CAS 328-93-8
Analyte or Component Names 2,5-Bis-(trifluoromethyl)aniline
Shipping Condition Room Temperature
Molecular Formula C8 H5 F6 N
Certifications/Compliance ISO 17034
Synonym 2,5-Bis(trifluoromethyl)aniline, 2,5-Bis(trifluoromethyl)benzenamine, 2,5-Di(trifluoromethyl)aniline
InChI Formula InChI=1S/C8H5F6N/c9-7(10,11)4-1-2-5(6(15)3-4)8(12,13)14/h1-3H,15H2
SMILES Nc1cc(ccc1C(F)(F)F)C(F)(F)F
IUPAC Name 2,5-bis(trifluoromethyl)aniline
Molecular Weight (g/mol) 229.12
Quantity 250 mg
Formula Weight 229.0326 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.