missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromohydroquinone, 97%

Product Code. 15461698
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15461698 5 g 5g
15451698 1 g 1g
15471698 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15461698 Supplier Thermo Scientific Alfa Aesar Supplier No. H55453.06
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 14753-51-6 | C6H4Br2O2 | 267.904 g/mol

It is an important pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important pharmaceutical intermediate.

Solubility
Soluble in methanol

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14753-51-6
Molecular Formula C6H4Br2O2
Molecular Weight (g/mol) 267.904
MDL Number MFCD00192664
InChI Key VALXCIRMSIFPFN-UHFFFAOYSA-N
PubChem CID 280945
IUPAC Name 2,5-dibromobenzene-1,4-diol
SMILES C1=C(C(=CC(=C1Br)O)Br)O

Specifications

Melting Point 191°C to 194°C
Quantity 5 g
Synonym 2,5-dibromohydroquinone, 2,5-dibromo-1,4-benzenediol, 2,5-dibromohydroquine, 1,4-benzenediol,2,5-dibromo, 2,5-dibromo-benzene-1,4-diol, 1,4-benzenediol, 2,5-dibromo, 2,5-dibromo-1,4-dihydroxybenzene
Solubility Information Soluble in methanol
Formula Weight 267.9
Percent Purity 97%
Chemical Name or Material 2,5-Dibromohydroquinone
compliance-icons
Product Identifier
  • 2,5-Dibromohydroquinone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2,5-Dibromohydroquinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.