missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Di-tert-butyl-4-methylphenol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Butylhydroxytoluene |
| CAS | 128-37-0 |
| Analyte or Component Names | Butylhydroxytoluene |
| Shipping Condition | +20°C |
| Molecular Formula | C15 H24 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Butylhydroxytoluene, 2,6-Bis(1,1-dimethylethyl)-4-methylphenol, 2,6-Bis(tert-butyl)-4-methylphenol, 2,6-Di(tert-butyl)hydroxytoluene, 2,6-Di-tert-butyl-4-cresol, 2,6-Di-tert-butyl-4-hydroxytoluene, 2,6-Di-tert-butyl-4-methyl-1-hydroxybenzene, 2,6-Di-tert-butyl-4-methylhydroxybenzene, 2,6-Di-tert-butyl-4-methylphenol, 2,6-Di-tert-butyl-p-cresol, 2,6-Di-tert-butyl-p-cresole, 2,6-Di-tert-butyl-p-methylphenol, 2,6-Di-tert-butylcresol, 2,6-Di-tert-butylmethylphenol, 2,6-tert-Butyl-4-methylphenol, 3,5-Di-tert-butyl-4-hydroxytoluene, 4-Hydroxy-3,5-di-tert-butyltoluene, 4-Methyl-2,6-bis(1,1-dimethylethyl)phenol, 4-Methyl-2,6-di-tert-butylphenol, AO 29, AO 4, AO 4K, AOX 4, AOX 4K, Advastab 401, Agidol, Agidol 1, Agidol 1A, Alkofen BP, Antage BHT, Antioxidant 264, Antioxidant 29, Antioxidant 30, Antioxidant 4, Antioxidant 4K, Antioxidant DBPC, Antioxidant KB, Antioxidant MPJ, Antioxidant T 501, Antox QT, B-NOX BHT-P, BAT, BHT, BHT 264, BHT Swanox, BHT-C, Buks, Butylated hydroxytoluene, CAO 1, CAO 3, Catalin CAO 3, Chemanox 11, DBPC, Dalpac, Deenax, Di-tert-Butyl-4-methylphenol, Di-tert-butyl-p-cresol, Di-tert-butylcresol, Dibunol, Dibutylated hydroxytoluene, Dibutylcresol, Dibutylhydroxytoluene, E 321, Impruvol, Ionol, Ionol 1, Ionol 330, Ionol BHT, Ionol CP, Ionol K, Ionole, Kerabit, Lowinox BHT, Lubrizol 817, NSC 6347, Naugard BHT, Nocrac 200, Nocrac 2000, Nonox TBC, P 21, P 21 (phenol), Parabar 441, Permanax BHT, Popol, Ralox BHT, Selosol H 633, Stavox, Sumilizer BHT, Sumilizer BHT-R, Sustane BHT, Swanox BHT, T 501, T 501 (phenol), Tenamene 3, Tenox BHT, Tonarol, Topanol, Topanol BHT, Topanol O, Topanol O-FG, Topanol OC, Topanol OL, Ultranox 226, Vanlube PC, Vanlube PCX, Vanox PC, Vianol, Vulkanox KB, Yoshinox BHT, o-Di-tert-butyl-p-methylphenol, BHT, 2,6-di-tert-butyl-4-methylphenol |
| InChI Formula | InChI=1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3 |
| SMILES | Cc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C |
| IUPAC Name | 2,6-ditert-butyl-4-methylphenol |
| Show More |
Product Title
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?