missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenol, 99%

Product Code. 11457037
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457037 500 g 500g
11437037 25 g 25g
11447037 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457037 Supplier Thermo Scientific Alfa Aesar Supplier No. A14411.36
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 87-65-0 | C6H4Cl2O | 163.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-65-0
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002176
InChI Key HOLHYSJJBXSLMV-UHFFFAOYSA-N
Synonym phenol, 2,6-dichloro, 2,6-dichlorfenol, 2,6-dichlorphenol, rcra waste number u082, 2,6-dichloro-phenol, 2,6-dichlorfenol czech, unii-q7e9k52w7e, 2,6 dichlorophenol, ccris 2511, 2,6-dcp
PubChem CID 6899
ChEBI CHEBI:28457
IUPAC Name 2,6-dichlorophenol
SMILES OC1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 65°C to 68°C
Boiling Point 218°C to 220°C
Odor Phenol-like
Quantity 500 g
UN Number UN2020
Beilstein 1447806
Merck Index 14,3073
Formula Weight 163
Percent Purity 99%
Chemical Name or Material 2,6-Dichlorophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.