missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenol, Mikromol™

Product Code. 17923666
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17923666 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17923666 Supplier Mikromol Supplier No. MM3572.040100
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,6-Dichlorophenol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,6-Dichlorophenol
CAS 87-65-0
Analyte or Component Names 2,6-Dichlorophenol
Shipping Condition Room Temperature
Molecular Formula C6 H4 Cl2 O
Certifications/Compliance ISO 9001
Synonym 2,6-Dichlorophenol, NSC 60647, Phloroglucinol Impurity I
InChI Formula InChI=1S/C6H4Cl2O/c7-4-2-1-3-5(8)6(4)9/h1-3,9H
SMILES Oc1c(Cl)cccc1Cl
IUPAC Name 2,6-dichlorophenol
Molecular Weight (g/mol) 163
Quantity 100 mg
Formula Weight 161.9639 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.