missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloropurine, Mikromol™

Product Code. 30049146
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049146 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049146 Supplier Mikromol Supplier No. MM0061.100025
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,6-Dichloropurine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,6-Dichloropurine
CAS 5451-40-1
Analyte or Component Names 2,6-Dichloropurine
Shipping Condition Room Temperature
Molecular Formula C5 H2 Cl2 N4
Certifications/Compliance ISO 9001
Synonym 2,6-Dichloropurine
InChI Formula InChI=1S/C5H2Cl2N4/c6-3-2-4(9-1-8-2)11-5(7)10-3/h1H,(H,8,9,10,11)
SMILES Clc1nc(Cl)c2nc[nH]c2n1
IUPAC Name 2,6-dichloro-9H-purine
Molecular Weight (g/mol) 189
Quantity 25 mg
Formula Weight 187.9657 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.