missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-4-nitrophenol, 98%

Product Code. p-7022612
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11432847

Brand: Thermo Scientific Alfa Aesar A15472.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2423-71-4 | C8H9NO3 | 167.164 g/mol

It was used as internal standard in determination of 4-nitrophenol and 3-methyl-4-nitrophenol in human urine by liquid chromatography combined with tandem mass spectrometry. ,6-Dimethyl-4-nitrophenol was used to study the disposition requirement for binding of p-nitrophenol and sodium p-nitrophenolate like substrates with cyclohexaamylaose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It was used as internal standard in determination of 4-nitrophenol and 3-methyl-4-nitrophenol in human urine by liquid chromatography combined with tandem mass spectrometry. ,6-Dimethyl-4-nitrophenol was used to study the disposition requirement for binding of p-nitrophenol and sodium p-nitrophenolate like substrates with cyclohexaamylaose.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2423-71-4
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00007339
InChI Key FNORUNUDZNWQFF-UHFFFAOYSA-N
PubChem CID 17030
IUPAC Name 2,6-dimethyl-4-nitrophenol
SMILES CC1=CC(=CC(=C1O)C)[N+](=O)[O-]

Specifications

Melting Point ∼168°C (decomposition)
Quantity 5 g
Beilstein 1873275
Synonym 4-nitro-2,6-xylenol, 2,6-xylenol, 4-nitro, 4-nitro-2,6-dimethylphenol, phenol, 2,6-dimethyl-4-nitro, phenol,2,6-dimethyl-4-nitro, acmc-1cljn, 2, 4-nitro, intermediates-zcf02651, 2,6-dimethyl4-nitrophenol, phenol,6-dimethyl-4-nitro
Solubility Information Insoluble in water.
Formula Weight 167.17
Percent Purity 98%
Chemical Name or Material 2,6-Dimethyl-4-nitrophenol
compliance-icons
Product Identifier
  • 2,6-Dimethyl-4-nitrophenol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 4-Nitro-2,6-xylenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.