missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylbenzamide, Thermo Scientific™

Product Code. 10099551
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10099551 1 g Amber Glass Bottle 1g
10671503 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10099551 Supplier Thermo Scientific Maybridge Supplier No. TL00794DA
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 39549-79-6
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
MDL Number MFCD00221474
InChI Key RUHKZVAPXHIWJH-UHFFFAOYSA-N
Synonym 2-amino-4-methyl-benzamide, benzamide, 2-amino-4-methyl, pubchem4030, 2-azanyl-4-methyl-benzamide, benzamide,2-amino-4-methyl, benzamide, 2-amino-4-methyl-9ci
PubChem CID 2801474
IUPAC Name 2-amino-4-methylbenzamide
SMILES CC1=CC(N)=C(C=C1)C(N)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 150.18
Percent Purity 97%
Chemical Name or Material 2-Amino-4-methylbenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.