missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylthiazole, TRC

Product Code. 30136088
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136088 5 g 5g
17914315 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136088 Supplier Toronto Research Chemicals Supplier No. TRCA6182355G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7305-71-7
Molecular Formula C4 H6 N2 S
Molecular Weight (g/mol) 114.17
Synonym 5-Methylthiazol-2-amine, Meloxicam Imp. B (EP)
IUPAC Name 5-methyl-1,3-thiazol-2-amine
SMILES Cc1cnc(N)s1

Specifications

Analyte or Component Names 5-Methylthiazol-2-amine
Shipping Condition Room Temperature
InChI Formula InChI=1S/C4H6N2S/c1-3-2-6-4(5)7-3/h2H,1H3,(H2,5,6)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 5 g
Formula Weight 114.0252
Chemical Name or Material 5-Methylthiazol-2-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.