missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloropurine, Mikromol™

Product Code. 17961846
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17961846 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17961846 Supplier Mikromol Supplier No. MM0620.200025
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Amino-6-chloropurine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-6-chloropurine
CAS 10310-21-1
Analyte or Component Names 2-Amino-6-chloropurine
Shipping Condition Room Temperature
Molecular Formula C5 H4 Cl N5
Certifications/Compliance ISO 17025
Synonym 9H-Purin-2-amine, 6-chloro-, 1H-Purin-2-amine, 6-chloro- (9CI), Purine, 2-amino-6-chloro- (6CI,7CI,8CI), 6-Chloro-9H-purin-2-amine, 2-Amino-6-chloro-9H-purine, 2-Amino-6-chloropurine, 6-Chloro-1H-purin-2-amine, 6-Chloro-2-aminopurine, 6-Chloro-2-purinamine, 6-Chloro-7H-purin-2-amine, 6-Chloro-7H-purin-2-ylamine, 6-Chloro-9H-purin-2-yl-2-amine, 6-Chloroguanine, NSC 29570, SKF-17831, 380U58, Nelarabine related, Famciclovir Related Compound F
InChI Formula InChI=1S/C5H4ClN5/c6-3-2-4(9-1-8-2)11-5(7)10-3/h1H,(H3,7,8,9,10,11)
SMILES Nc1nc(Cl)c2nc[nH]c2n1
IUPAC Name 6-chloro-9H-purin-2-amine
Molecular Weight (g/mol) 169.57
Quantity 25 mg
Formula Weight 169.0155 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.