missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethanol (Ethanolamine), Mikromol™

Product Code. 18062970
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18062970 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18062970 Supplier Mikromol Supplier No. MM1204.01
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Aminoethanol (Ethanolamine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethanolamine
CAS 141-43-5
Analyte or Component Names Ethanolamine
Shipping Condition Room Temperature
Molecular Formula C2 H7 N O
Certifications/Compliance ISO 17025
Synonym C2H7NO, Trolamine Imp. A (EP), 2-Aminoethanol, 1-Amino-2-hydroxyethane, 2-Amino-1-ethanol, 2-Aminoethyl alcohol, 2-Ethanolamine, 2-Hydroxyethanamine, 2-Hydroxyethylamine, Aminoethanol, Colamine, ETA, Envision Conditioner PDD 9020, Ethanolamine, Ethylolamine, Glycinol, Kolamin, MEA, MEA (alcohol), MEA 90, MEA-LCI, Mealan, Monoethanolamine, Olamine, R 100, R 100 (amine), Seramine, beta-Aminoethanol, beta-Aminoethyl alcohol, beta-Ethanolamine, beta-Hydroxyethylamine, Ifosfamide Impurity D, Trolamine Impurity A, 2-Aminoethanol, 1,10-Decanedicarboxylic Acid, 1,10-Dicarboxydecane, 1,12-Dodecanedioic Acid, Corfree M 2, Decamethylenedicarboxylic Acid, NSC 400242, SL-AH, n-Dodecane-α,ω-dioic Acid, n-Dodecanedioic Acid
InChI Formula InChI=1S/C2H7NO/c3-1-2-4/h4H,1-3H2
SMILES NCCO
IUPAC Name 2-aminoethanol
Molecular Weight (g/mol) 61.08
Quantity 100 mg
Formula Weight 61.0528 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.