missing translation for 'onlineSavingsMsg'
Learn More

2-[Benzyl(1,1-dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone Hydrochloride, Mikromol™

Product Code. 30049960
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049960 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049960 Supplier Mikromol Supplier No. MM0381.07
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-[Benzyl(1,1-dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[Benzyl(1,1-dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone Hydrochloride
CAS 24085-08-3
Analyte or Component Names 2-[Benzyl(1,1-dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone Hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C20 H25 N O3 . Cl H
Certifications/Compliance ISO 9001
Synonym Salbutamol Sulphate Imp. G (EP), Salbutamol Imp. G (EP), 2-[Benzyl(1,1-dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone Hydrochloride, Salbutamol Sulphate Imp. G (EP) as Hydrochloride, Salbutamol Sulfate Impurity G, Salbutamol Impurity G
InChI Formula InChI=1S/C20H25NO3.ClH/c1-20(2,3)21(12-15-7-5-4-6-8-15)13-19(24)16-9-10-18(23)17(11-16)14-22;/h4-11,22-23H,12-14H2,1-3H3;1H
SMILES Cl.CC(C)(C)N(CC(=O)c1ccc(O)c(CO)c1)Cc2ccccc2
IUPAC Name 2-[benzyl(tert-butyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone;hydrochloride
Molecular Weight (g/mol) 363.88
Quantity 50 mg
Formula Weight 363.1601 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.