missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-(4-methylphenyl)ethanone, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
2-Bromo-1-(4-methylphenyl)ethanone
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-4'-methylacetophenone |
| CAS | 619-41-0 |
| Analyte or Component Names | 2-Bromo-4'-methylacetophenone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H9 Br O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ethanone, 2-bromo-1-(4-methylphenyl)-, Acetophenone, 2-bromo-4'-methyl- (6CI,7CI,8CI), 2-Bromo-1-(4-methylphenyl)ethanone, 2-Bromo-1-(4-methylphenyl)ethan-1-one, 2-Bromo-1-(4-tolyl)ethanone, 2-Bromo-1-(p-methylphenyl)ethanone, 2-Bromo-1-(p-tolyl)ethanone, 2-Bromo-4'-methylacetophenone, 2-Bromo-p-methylacetophenone, 4-Methylphenacyl bromide, 4'-Methyl-2-bromoacetophenone, Bromomethyl 4-methylphenyl ketone, Bromomethyl p-tolyl ketone, NSC 63192, p-Methyl-Ω-bromoacetophenone, p-Methylphenacyl bromide, Ω-Bromo-p-methylacetophenone, 4-Methyl-alpha-bromoacetophenone, p-Methyl-alpha-bromoacetophenone, p-Methyl-omega-bromoacetophenone, alpha-Bromo-4'-methylacetophenone, alpha-Bromo-p-methylacetophenone, omega-Bromo-p-methylacetophenone |
| InChI Formula | InChI=1S/C9H9BrO/c1-7-2-4-8(5-3-7)9(11)6-10/h2-5H,6H2,1H3 |
| SMILES | Cc1ccc(cc1)C(=O)CBr |
| IUPAC Name | 2-bromo-1-(4-methylphenyl)ethanone |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction