missing translation for 'onlineSavingsMsg'
Learn More

2-bromo-5-fluorophenyl isothiocyanate, Thermo Scientific™

Product Code. 10205402
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10205402 1 g 1g
10312315 5 g 5g
10260983 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10205402 Supplier Thermo Scientific Maybridge Supplier No. SB01713DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Identificatori chimici

CAS 175205-35-3
Molecular Formula C7H3BrFNS
Molecular Weight (g/mol) 232.07
MDL Number MFCD00673194
InChI Key SSLVXFSUADEZBQ-UHFFFAOYSA-N
Synonym 2-bromo-4-fluorophenyl isothiocyanate, 2-bromo-4-fluorophenylisothiocyanate, pubchem5018, acmc-20ao2g, 2-bromo-4-fluorobenzenisothiocyanate, benzene,1-bromo-4-fluoro-2-isothiocyanato, 2-bromo-4-fluorophenyl isothiocyanate, tech., benzene, 2-bromo-4-fluoro-1-isothiocyanato, 2-bromanyl-4-fluoranyl-1-isothiocyanato-benzene, benzene, 2-bromo-4-fluoro-1-isothiocyanato-9ci
PubChem CID 2736332
IUPAC Name 2-bromo-4-fluoro-1-isothiocyanatobenzene
SMILES FC1=CC=C(N=C=S)C(Br)=C1

Specifica

Quantity 1 g
Formula Weight 232.08
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Bromo-5-fluorophenyl isothiocyanate
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.