missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-methoxyphenol, 98+%

Product Code. p-7028298
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11461070

Brand: Thermo Scientific Alfa Aesar B25535.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28165-49-3 | C7H7BrO2 | 203.035 g/mol

2-Bromo-6-methoxyphenol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-6-methoxyphenol is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28165-49-3
Molecular Formula C7H7BrO2
Molecular Weight (g/mol) 203.035
MDL Number MFCD08146628
InChI Key WEUFQISIJPSTBM-UHFFFAOYSA-N
PubChem CID 11019958
IUPAC Name 2-bromo-6-methoxyphenol
SMILES COC1=C(C(=CC=C1)Br)O

Specifications

Melting Point 62°C to 65°C
Quantity 1 g
Synonym 2-bromo-6-methoxy-phenol, phenol, 2-bromo-6-methoxy, bromhydroxyanisol, 6-bromoguaiacol, 3-bromo-2-hydroxyanisole, acmc-1ch0f, phenol,2-bromo-6-methoxy, 2-bromanyl-6-methoxy-phenol, ksc560s4p
Solubility Information Slightly soluble in water.
Formula Weight 203.04
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-6-methoxyphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.