missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzamide, 98%

Product Code. 11433897
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433897 5 g 5g
11443897 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11433897 Supplier Thermo Scientific Alfa Aesar Supplier No. A18331.06
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4001-73-4 | C7H6BrNO | 200.035 g/mol

2-Bromobenzamide has been used in microwave assisted one-pot synthesis of substituted 3-(phenylmethylene)isoindolin-1-ones, palladium-catalyzed synthesis of phenanthridinones and in the synthesis of new water-soluble iminophosphorane ligand.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzamide has been used in microwave assisted one-pot synthesis of substituted 3-(phenylmethylene)isoindolin-1-ones, palladium-catalyzed synthesis of phenanthridinones and in the synthesis of new water-soluble iminophosphorane ligand.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4001-73-4
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.035
MDL Number MFCD00007969
InChI Key NHNAEZDWNCRWRW-UHFFFAOYSA-N
Synonym benzamide, 2-bromo, o-bromobenzamide, benzamide, o-bromo, 2-bromo-benzamide, 2-bromobenzamide, bromobenzamide, pubchem3733, maybridge1_005867, acmc-209j9x
PubChem CID 77616
IUPAC Name 2-bromobenzamide
SMILES C1=CC=C(C(=C1)C(=O)N)Br

Specifications

Melting Point 160°C to 162°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 200.03
Percent Purity 98%
Chemical Name or Material 2-Bromobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.