missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenylacetonitrile, 97%

Product Code. 11432000
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432000 5 g 5g
11442000 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11432000 Supplier Thermo Scientific Alfa Aesar Supplier No. A17548.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19472-74-3 | C8H6BrN | 196.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19472-74-3
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD00001896
InChI Key BVCOJESIQPNOIF-UHFFFAOYSA-N
Synonym 2-bromophenylacetonitrile, 2-bromobenzyl cyanide, 2-2-bromophenyl acetonitrile, o-bromobenzyl cyanide, 2-bromophenyl acetonitrile, o-bromophenylacetonitrile, benzeneacetonitrile, bromo, 2-2-bromophenyl ethanenitrile, benzeneacetonitrile, 2-bromo, 2-bromo-phenyl-acetonitrile
PubChem CID 29625
IUPAC Name 2-(2-bromophenyl)acetonitrile
SMILES BrC1=CC=CC=C1CC#N

Specifications

Melting Point 19°C
Boiling Point 252°C
Flash Point 113°C (235°F)
Odor Pungent
Quantity 5 g
UN Number UN1694
Beilstein 2043684
Formula Weight 196.05
Percent Purity 97%
Chemical Name or Material 2-Bromophenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.