missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiophenol, 97%

Product Code. 11359899
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359899 5 g 5g
11369899 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359899 Supplier Thermo Scientific Alfa Aesar Supplier No. B25537.06
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 6320-02-1 | C6H5ClS | 144.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6320-02-1
MDL Number MFCD00004827

Specifications

Density 1.604
Boiling Point 114°C to 116°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Molecular Formula C6H5ClS
Refractive Index 1.635
Quantity 5 g
UN Number UN2922
Beilstein 2039769
Sensitivity Air Sensitive
Synonym 2-chlorothiophenol, o-chlorothiophenol, benzenethiol, 2-chloro, 1-chloro-2-mercaptobenzene, o-chlorobenzenethiol, o-chloromercaptobenzene, chlorothiophenol, benzenethiol, o-chloro, 2-chlorobenzene-1-thiol, 2-chloro thiophenol
InChI Key PWOBDMNCYMQTCE-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)S)Cl
IUPAC Name 2-chlorobenzenethiol
Molecular Weight (g/mol) 144.62
PubChem CID 80599
Formula Weight 189.08
Percent Purity 97%
Chemical Name or Material 2-Bromothiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.