missing translation for 'onlineSavingsMsg'
Learn More

2-Butyl-4-chloro-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-Imidazole-5-carboxaldehyde, Mikromol™

Product Code. 30051434
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051434 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051434 Supplier Mikromol Supplier No. MM0168.310025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Butyl-4-chloro-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-Imidazole-5-carboxaldehyde
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Trityl Losartan Carboxaldehyde
CAS 133910-00-6
Analyte or Component Names N-Trityl Losartan Carboxaldehyde
Shipping Condition Room Temperature
Molecular Formula C41H35ClN6O
Certifications/Compliance ISO 9001
Synonym 2-Butyl-4-chloro-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-Imidazole-5-carboxaldehyde, N-Trityl Losartan Carboxaldehyde
InChI Formula InChI=1S/C41H35ClN6O/c1-2-3-23-38-43-39(42)37(29-49)47(38)28-30-24-26-31(27-25-30)35-21-13-14-22-36(35)40-44-46-48(45-40)41(32-15-7-4-8-16-32,33-17-9-5-10-18-33)34-19-11-6-12-20-34/h4-22,24-27,29H,2-3,23,28H2,1H3
SMILES CCCCc1nc(Cl)c(C=O)n1Cc2ccc(cc2)c3ccccc3c4nnn(n4)C(c5ccccc5)(c6ccccc6)c7ccccc7
IUPAC Name 2-butyl-5-chloro-3-[[4-[2-(2-trityltetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carbaldehyde
Molecular Weight (g/mol) 663.21
Quantity 25 mg
Formula Weight 662.2561 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.