missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzaldehyde, Mikromol™

Product Code. 30051068
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051068 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051068 Supplier Mikromol Supplier No. MM0383.160500
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Chlorobenzaldehyde
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chlorobenzaldehyde
CAS 89-98-5
Analyte or Component Names 2-Chlorobenzaldehyde
Shipping Condition Ice pack (-20°C)
Molecular Formula C7 H5 Cl O
Certifications/Compliance ISO 9001
Synonym Benzaldehyde, o-chloro- (8CI), 2-Chlorobenzaldehyde, 2-Chlorobenzoic aldehyde, 2-Chlorophenylcarboxaldehyde, NSC 15347, o-Chlorobenzaldehyde, o-Chlorobenzenecarboxaldehyde, o-Chloroformylbenzene
InChI Formula InChI=1S/C7H5ClO/c8-7-4-2-1-3-6(7)5-9/h1-5H
SMILES Clc1ccccc1C=O
IUPAC Name 2-chlorobenzaldehyde
Molecular Weight (g/mol) 140.567
Quantity 500 mg
Formula Weight 140.0029 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.