missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl chloride, 98+%

Product Code. 11379639
Click to view available options
Quantity:
250 g
500 g
2500 g
Unit Size:
2500g
250g
500g
This item is not returnable. View return policy

Product Code. 11379639

Brand: Thermo Scientific Alfa Aesar A11900.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 611-19-8 | C7H6Cl2 | 161.025 g/mol

2-Chlorobenzyl chloride was used as a sensory irritant to evaluate the characteristic modifications of the normal breathing pattern of exposed mice. It is used as an intermediate for organic chemicals, pharmaceuticals, and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzyl chloride was used as a sensory irritant to evaluate the characteristic modifications of the normal breathing pattern of exposed mice. It is used as an intermediate for organic chemicals, pharmaceuticals, and dyes.

Solubility
Not miscible in water.

Notes
Store away from strong bases and oxidizing agents. Keep away from heat. Incompatible with Alcohols, Amines, Metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-19-8
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000893
InChI Key BASMANVIUSSIIM-UHFFFAOYSA-N
Synonym 2-chlorobenzyl chloride, 1-chloro-2-chloromethyl benzene, 2-chlorobenzylchloride, o-chlorobenzyl chloride, alpha,2-dichlorotoluene, benzene, 1-chloro-2-chloromethyl, alpha,o-dichlorotoluene, ar,alpha-dichlorotoluene, ortho-alpha-dichlorotoluene, unii-h58di83rmd
PubChem CID 11906
IUPAC Name 1-chloro-2-(chloromethyl)benzene
SMILES C1=CC=C(C(=C1)CCl)Cl

Specifications

Melting Point -13°C
Boiling Point 213°C to 214°C
Flash Point 109°C (228°F)
Odor Pungent
Quantity 250 g
UN Number UN2235
Beilstein 471700
Formula Weight 161.03
Percent Purity ≥98%
Chemical Name or Material 2-Chlorobenzyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.