missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl p-tolyl sulfone, 98%, Thermo Scientific™

Product Code. p-7044206
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11367365

Brand: Thermo Scientific Alfa Aesar L05256.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22381-53-9
Molecular Formula C9H11ClO2S
Molecular Weight (g/mol) 218.695
MDL Number MFCD00051607
InChI Key KUAGAJLNIOIGRN-UHFFFAOYSA-N
Synonym sulfone, 2-chloroethyl p-tolyl, 2-chloroethyl p-tolyl sulfone, 1-2-chloroethanesulfonyl-4-methylbenzene, 1-2-chloroethyl sulfonyl-4-methylbenzene, 2-chloro-1-4-methylphenyl sulfonyl ethane, p-toluene 2-chloroethyl sulphone, 2-chloroethylp-tolylsulfone, benzene, 1-2-chloroethyl sulfonyl-4-methyl, 2-chloroethyl p-tolyl sulphone
PubChem CID 188162
IUPAC Name 1-(2-chloroethylsulfonyl)-4-methylbenzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)CCCl

Specifications

Melting Point 77°C to 81°C
Quantity 1 g
Beilstein 1107627
Formula Weight 218.7
Percent Purity 98%
Chemical Name or Material 2-Chloroethyl p-tolyl sulfone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.