missing translation for 'onlineSavingsMsg'
Learn More

2-Chloromethyl-3,5-dimethyl-4-methoxypyridine Hydrochloride, Mikromol™

Product Code. 30462241
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462241 4 x 25 mg 100mg
30052289 25 mg 25mg
30052651 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30462241 Supplier Mikromol Supplier No. MM0095.5500254AMP
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Chloromethyl-3,5-dimethyl-4-methoxypyridine Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloromethyl-3,5-dimethyl-4-methoxypyridine Hydrochloride
CAS 86604-75-3
Analyte or Component Names 2-Chloromethyl-3,5-dimethyl-4-methoxypyridine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C9 H12 Cl N O . Cl H
Certifications/Compliance ISO 17025
Synonym 2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride (1:1), Pyridine, 2-(chloromethyl)-4-methoxy-3,5-dimethyl-, hydrochloride (9CI), 2-(Chloromethyl)-3,5-dimethyl-4-methoxypyridine hydrochloride, 2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride, 3,5-Dimethyl-2-chloromethyl-4-methoxypyridine hydrochloride, Cc-993
InChI Formula InChI=1S/C9H12ClNO.ClH/c1-6-5-11-8(4-10)7(2)9(6)12-3;/h5H,4H2,1-3H3;1H
Solution Type Neat
SMILES Cl.COc1c(C)cnc(CCl)c1C
IUPAC Name 2-(chloromethyl)-4-methoxy-3,5-dimethylpyridine;hydrochloride
Molecular Weight (g/mol) 222.11
Quantity 4 x 25 mg
Formula Weight 221.0374 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.