missing translation for 'onlineSavingsMsg'
Learn More

(2-Chlorophenyl)diphenylmethanol, Mikromol™

Product Code. 30462926
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462926 4 x 25 mg -
30051691 25 mg 25mg
17963786 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30462926 Supplier Mikromol Supplier No. MM0015.0100254AMP
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2-Chlorophenyl)diphenylmethanol, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2-Chlorophenyl)diphenylmethanol
CAS 66774-02-5
Analyte or Component Names (2-Chlorophenyl)diphenylmethanol
Shipping Condition Room Temperature
Molecular Formula C19 H15 Cl O
Certifications/Compliance ISO 17025
Synonym Clotrimazole Imp. A (EP), Clotrimazole USP Related Compound A, Clotrimazole USP RC A, (2-Chlorophenyl)diphenylmethanol, Benzenemethanol, 2-chloro-alpha,alpha-diphenyl-, Methanol, (o-chlorophenyl)diphenyl- (6CI), NSC 170882, TRAM 3, o-Chlorophenyldiphenylmethanol, USP Clotrimazole Related Compound A, Ph Eur Clotrimazole Impurity A, Clotrimazole Related Compound A, Clotrimazole Impurity A
InChI Formula InChI=1S/C19H15ClO/c20-18-14-8-7-13-17(18)19(21,15-9-3-1-4-10-15)16-11-5-2-6-12-16/h1-14,21H
Solution Type Neat
SMILES OC(c1ccccc1)(c2ccccc2)c3ccccc3Cl
IUPAC Name (2-chlorophenyl)-diphenylmethanol
Molecular Weight (g/mol) 294.77
Quantity 4 x 25 mg
Formula Weight 294.0811 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.