missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxy-1-naphthoic acid, 98%, Thermo Scientific™

Product Code. 11495287
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495287 25 g 25g
11405297 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495287 Supplier Thermo Scientific Alfa Aesar Supplier No. B22665.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2224-00-2
Molecular Formula C13H11O3
Molecular Weight (g/mol) 215.23
MDL Number MFCD00004008
InChI Key MYFBSSDLYGWAHH-UHFFFAOYSA-M
Synonym 2-ethoxy-1-naphthoic acid, 2-ethoxynaphthoic acid, 1-naphthalenecarboxylic acid, 2-ethoxy, 2-ethoxy-1-naphthoicacid, 2-ethoxynaphthalenecarboxylic acid, acmc-209ftv, rarechem al be 0566, timtec-bb sbb005771, 2-ethoxynaphtoic acid
PubChem CID 75210
IUPAC Name 2-ethoxynaphthalene-1-carboxylic acid
SMILES CCOC1=CC=C2C=CC=CC2=C1C([O-])=O

Specifications

Melting Point 143°C to 145°C
Quantity 25 g
Beilstein 2695612
Formula Weight 216.24
Percent Purity 98%
Chemical Name or Material 2-Ethoxy-1-naphthoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.