accessibility menu, dialog, popup

UserName

H3COOO

CAS RN 2224-00-2

IUPAC Name: 2-ethoxynaphthalene-1-carboxylate
Synonyms: 2-ethoxynaphthalene-1-carboxylate
Molecular Weight (g/mol): 215.23
Molecular Formula: C13H11O3
InChi Key: MYFBSSDLYGWAHH-UHFFFAOYSA-M
SMILES: CCOC1=CC=C2C=CC=CC2=C1C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results