missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxynaphthoic acid, 98%, Thermo Scientific™

Product Code. 10522061 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10522061 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10522061 Supplier Thermo Scientific Acros Supplier No. 243000100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2224-00-2
Molecular Formula C13H11O3
Molecular Weight (g/mol) 215.23
MDL Number MFCD00004008
InChI Key MYFBSSDLYGWAHH-UHFFFAOYSA-M
Synonym 2-ethoxy-1-naphthoic acid, 2-ethoxynaphthoic acid, 1-naphthalenecarboxylic acid, 2-ethoxy, 2-ethoxy-1-naphthoicacid, 2-ethoxynaphthalenecarboxylic acid, acmc-209ftv, rarechem al be 0566, timtec-bb sbb005771, 2-ethoxynaphtoic acid
PubChem CID 75210
SMILES CCOC1=CC=C2C=CC=CC2=C1C([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 141°C to 145°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 10 g
IUPAC Name 2-ethoxynaphthalene-1-carboxylic acid
Formula Weight 216.24
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Ethoxynaphthoic acid, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.