missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl salicylate, Thermo Scientific™

Product Code. 17342501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17342501 25 mL 25mL
17352501 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17342501 Supplier Thermo Scientific Acros Supplier No. 468830250
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 118-60-5
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.34
MDL Number 00053300
InChI Key FMRHJJZUHUTGKE-UHFFFAOYNA-N
IUPAC Name 2-ethylhexyl 2-hydroxybenzoate
SMILES CCCCC(CC)COC(=O)C1=CC=CC=C1O

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Density 1.02
Flash Point 162°C
Physical Form Liquid
Color Colorless to Light Yellow
Melting Point -20°C
Boiling Point 190°C (21 mmHG)
Solubility Insoluble in water
Quantity 25 mL
Formula Weight 250.34
Chemical Name or Material 2-Ethylhexyl salicylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.