missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-methylbenzyl bromide, 97%

Product Code. p-7025176
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478907 1 g 1g
11488907 5 g 5g
11498907 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478907 Supplier Thermo Scientific Alfa Aesar Supplier No. B20538.03
Add to basket
This item is not returnable. View return policy

CAS: 151412-12-3 | C8H8BrF | 203.054 g/mol

2-Fluoro-3-methylbenzyl bromide, is used as a chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-3-methylbenzyl bromide, is used as a chemical and organic intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151412-12-3
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.054
MDL Number MFCD00042470
InChI Key DCGWFNZSJBIAKE-UHFFFAOYSA-N
Synonym 2-fluoro-3-methylbenzyl bromide, 1-bromomethyl-2-fluoro-3-methylbenzene, benzene,1-bromomethyl-2-fluoro-3-methyl, acmc-20anvi, 2-fluoro-3-methylbenzylbromide, 2-fluoro-3-methyl-benzylbromide, 2-fluoro-3-methyl benzyl bromide, 3-bromomethyl-2-fluoro-1-methylbenzene, 1-bromomethyl-2-fluoro-3-methylbenzene #
PubChem CID 519062
IUPAC Name 1-(bromomethyl)-2-fluoro-3-methylbenzene
SMILES CC1=C(C(=CC=C1)CBr)F

Specifications

Boiling Point 105°C to 106°C (10 mmHg)
Refractive Index 1.548
Quantity 1 g
UN Number UN3265
Beilstein 7473765
Solubility Information Insoluble in water.
Formula Weight 203.06
Percent Purity 97%
Chemical Name or Material 2-Fluoro-3-methylbenzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.