missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-iodotoluene, 99%, Thermo Scientific™

Product Code. p-7020646
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11440563

Brand: Thermo Scientific Alfa Aesar A12679.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39998-81-7
Molecular Formula C7H6FI
Molecular Weight (g/mol) 236.028
MDL Number MFCD00001045
InChI Key QZLWTFTXGKKCHZ-UHFFFAOYSA-N
Synonym 2-fluoro-4-iodotoluene, 2-fluoro-4-iodo-1-methyl-benzene, benzene, 2-fluoro-4-iodo-1-methyl, 2-fluor-4-iod-1-methylbenzol, pubchem1618, 2-fluoro-4-iodo-toluene, acmc-209j9t, 3-fluoro-4-methyl-iodobenzene, buttpark 50\01-77, #
PubChem CID 123492
IUPAC Name 2-fluoro-4-iodo-1-methylbenzene
SMILES CC1=C(C=C(C=C1)I)F

Specifications

Melting Point 22°C to 24°C
Boiling Point 206°C to 208°C
Flash Point 79°C (174°F)
Refractive Index 1.578
Quantity 5 g
Beilstein 2432998
Sensitivity Light sensitive
Formula Weight 236.03
Percent Purity 99%
Chemical Name or Material 2-Fluoro-4-iodotoluene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.