missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzothiazole, 99%

Product Code. 11314236
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314236 250 mg 250mg
11394226 1 g 1g
11304236 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11314236 Supplier Thermo Scientific Alfa Aesar Supplier No. L19316.MD
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1123-98-4 | C7H4FNS | 153.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1123-98-4
Molecular Formula C7H4FNS
Molecular Weight (g/mol) 153.174
MDL Number MFCD00047254
InChI Key QVWCHVAUHZEAAT-UHFFFAOYSA-N
Synonym 2-fluorobenzothiazole, 2-fluorobenzo d thiazole, 2-benzothiazolyl fluoride, benzothiazole, 2-fluoro, 2-fluoro-benzothiazole, fluorobenzothiazole, acmc-2099en, #
PubChem CID 70753
IUPAC Name 2-fluoro-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)F

Specifications

Density 1.33
Boiling Point 97°C to 98°C (16 mmHg)
Odor Characteristic
Refractive Index 1.583
Quantity 250 mg
UN Number UN2810
Beilstein 971503
Formula Weight 153.18
Percent Purity 99%
Chemical Name or Material 2-Fluorobenzothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.