missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methoxybenzophenone, 98+%

Product Code. 11473238
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473238 50 g 50g
11483238 250 g 250g
11463238 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473238 Supplier Thermo Scientific Alfa Aesar Supplier No. A17662.18
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 131-57-7 | C14H12O3 | 228.247 g/mol

2-Hydroxy-4-methoxybenzophenone is used as a broad-band filter in sunscreens cosmetic products in order to protect the skin from the deleterious effects of the sun. It acts as a reagent in the gravimetric determination of copper(II). Further, it is used in the preparation of poly[(2-hydroxy-4-methoxybenzophenone) propylene], which is a polymeric ligand useful for the synthesis of metal/ligand polychelates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-4-methoxybenzophenone is used as a broad-band filter in sunscreens cosmetic products in order to protect the skin from the deleterious effects of the sun. It acts as a reagent in the gravimetric determination of copper(II). Further, it is used in the preparation of poly[(2-hydroxy-4-methoxybenzophenone) propylene], which is a polymeric ligand useful for the synthesis of metal/ligand polychelates.

Solubility
Insoluble in water.

Notes
Store in a cool place. Incompatible with bases, acid chlorides, acid anhydrides and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-57-7
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD00008387
InChI Key DXGLGDHPHMLXJC-UHFFFAOYSA-N
Synonym oxybenzone, 2-hydroxy-4-methoxybenzophenone, benzophenone-3, oxybenzon, 4-methoxy-2-hydroxybenzophenone, 2-benzoyl-5-methoxyphenol, oxibenzona, oxybenzonum, 2-hydroxy-4-methoxyphenyl phenyl methanone, anuvex
PubChem CID 4632
ChEBI CHEBI:34283
IUPAC Name (2-hydroxy-4-methoxyphenyl)-phenylmethanone
SMILES COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O

Specifications

Melting Point 62°C to 65°C
Density 1.3
Boiling Point ∼150°C (5mmHg)
Flash Point 216°C (420°F)
Quantity 50 g
Beilstein 1913145
Merck Index 14,6954
Solubility Information Insoluble in water.
Formula Weight 228.25
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxy-4-methoxybenzophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.