accessibility menu, dialog, popup

UserName

H3COHOO

CAS RN 131-57-7

IUPAC Name: 2-benzoyl-5-methoxyphenol
Synonyms: oxybenzone oxybenzone (substance) oxibenzona [inn-spanish] chimassorb 90 oxybenzone preparation solbar spectra-sorb UV 9 person & covey brand of oxybenzone MOB MOD
Molecular Weight (g/mol): 228.25
Molecular Formula: C14H12O3
InChi Key: DXGLGDHPHMLXJC-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(=O)C2=CC=CC=C2)C(O)=C1
Molecular Weight (g/mol) 
  • (6)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)