missing translation for 'onlineSavingsMsg'
Learn More

2-Indanylacetic acid, 99%

Product Code. 11332946
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332946 1 g 1g
11342946 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332946 Supplier Thermo Scientific Alfa Aesar Supplier No. L10700.03
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 37868-26-1 | C11H12O2 | 176.22 g/mol

2-Indanylacetic acid is the starting material in the synthesis of pentacylic core of (+)-Salvileucalin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Indanylacetic acid is the starting material in the synthesis of pentacylic core of (+)-Salvileucalin.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37868-26-1
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.22
MDL Number MFCD00082653
InChI Key TULDPXYHBFBRGW-UHFFFAOYSA-N
Synonym 2-2,3-dihydro-1h-inden-2-yl acetic acid, indan-2-yl-acetic acid, 1h-indene-2-acetic acid, 2,3-dihydro, 2,3-dihydro-1h-inden-2-ylacetic acid, 2,3-dihydro-1h-indene-2-acetic acid, 2-indaneacetic acid, indan-2-acetic acid, pubchem16311, indan-2-yl acetic acid
PubChem CID 5174955
IUPAC Name 2-(2,3-dihydro-1H-inden-2-yl)acetic acid
SMILES OC(=O)CC1CC2=CC=CC=C2C1

Specifications

Melting Point 91°C to 93°C
Quantity 1 g
Beilstein 2616409
Solubility Information Insoluble in water.
Formula Weight 176.22
Percent Purity 99%
Chemical Name or Material 2-Indanylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.