missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Isopropoxyethyl 4-[[(2RS)-2-Hydroxy-3-(isopropylamino)propyl]oxy]benzoate (Bisoprolol Ester), Mikromol™
Description
2-Isopropoxyethyl 4-[[(2RS)-2-Hydroxy-3-(isopropylamino)propyl]oxy]benzoate (Bisoprolol Ester)
Specifications
Specifications
| Chemical Name or Material | 2-Isopropoxyethyl 4-([(2RS)-2-Hydroxy-3-(isopropylamino)propyl]oxy)benzoate (Bisoprolol Ester) |
| CAS | 864544-37-6 |
| Analyte or Component Names | 2-Isopropoxyethyl 4-([(2RS)-2-Hydroxy-3-(isopropylamino)propyl]oxy)benzoate (Bisoprolol Ester) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H29 N O5 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Bisoprolol Fumarate Imp. K (EP), Bisoprolol Imp. K (EP), 2-Isopropoxyethyl 4-[[(2RS)-2-Hydroxy-3-(isopropylamino)propyl]oxy]benzoate, Bisoprolol Ester, Bisoprolol Fumarate Impurity K, Bisoprolol Impurity K |
| InChI Formula | InChI=1S/C18H29NO5/c1-13(2)19-11-16(20)12-24-17-7-5-15(6-8-17)18(21)23-10-9-22-14(3)4/h5-8,13-14,16,19-20H,9-12H2,1-4H3 |
| SMILES | CC(C)NCC(O)COc1ccc(cc1)C(=O)OCCOC(C)C |
| IUPAC Name | 2-propan-2-yloxyethyl 4-[2-hydroxy-3-(propan-2-ylamino)propoxy]benzoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?