missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylbenzeneboronic acid, 97%

Product Code. 11304556
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304556 1 g 1g
11324556 250 mg 250mg
11314556 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11304556 Supplier Thermo Scientific Alfa Aesar Supplier No. L20110.03
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 89787-12-2 | C9H13BO2 | 164.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89787-12-2
Molecular Formula C9H13BO2
Molecular Weight (g/mol) 164.01
MDL Number MFCD03411937
InChI Key KTZUVUWIBZMHMC-UHFFFAOYSA-N
Synonym 2-isopropylphenylboronic acid, 2-isopropylphenyl boronic acid, 2-isopropylbenzeneboronic acid, 2-cumylboronic acid, 2-propan-2-ylphenyl boronic acid, o-isopropylphenylboronic acid, 2-isopropylphenboronic acid, 2-isopropylphenyboronic acid, 2-propan-2-yl phenyl boronic acid, boronic acid, 2-1-methylethyl phenyl
PubChem CID 2773477
IUPAC Name (2-propan-2-ylphenyl)boronic acid
SMILES CC(C)C1=CC=CC=C1B(O)O

Specifications

Melting Point 85°C to 86°C
Quantity 1 g
Beilstein 3090298
Formula Weight 164.01
Percent Purity 97%
Chemical Name or Material 2-Isopropylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.