missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzoic acid, 98+%

Product Code. 11495613
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495613 500 g 500g
11485613 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495613 Supplier Thermo Scientific Alfa Aesar Supplier No. A13513.36
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 579-75-9 | C8H8O3 | 152.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 579-75-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002431
InChI Key ILUJQPXNXACGAN-UHFFFAOYSA-N
Synonym o-anisic acid, o-methoxybenzoic acid, 2-anisic acid, o-methylsalicylic acid, benzoic acid, 2-methoxy, salicylic acid methyl ether, 2-methoxy-benzoic acid, 2-methoxybenzoicacid, o-methoxy benzoic acid, ortho-methoxybenzoic acid
PubChem CID 11370
ChEBI CHEBI:421840
IUPAC Name 2-methoxybenzoic acid
SMILES COC1=CC=CC=C1C(=O)O

Specifications

Melting Point 99°C to 103°C
Odor Odorless
Quantity 500 g
Beilstein 509929
Formula Weight 152.15
Percent Purity ≥98%
Chemical Name or Material 2-Methoxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.