missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxynaphthalene(Nerolin), Mikromol™

Product Code. 17932666
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17932666 25 mg 25mg
17910776 100 mg 100mg
30461423 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17932666 Supplier Mikromol Supplier No. MM0152.080025
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Methoxynaphthalene(Nerolin)
TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material 2-Methoxynaphthalene(Nerolin)
CAS 93-04-9
Analyte or Component Names 2-Methoxynaphthalene(Nerolin)
Shipping Condition Room Temperature
Molecular Formula C11 H10 O
Certifications/Compliance ISO 17025
Synonym 2-Methoxynaphthalene(Nerolin), Naproxen Imp. M (EP), Naproxen Impurity M
InChI Formula InChI=1S/C11H10O/c1-12-11-7-6-9-4-2-3-5-10(9)8-11/h2-8H,1H3
SMILES COc1ccc2ccccc2c1
IUPAC Name 2-methoxynaphthalene
Molecular Weight (g/mol) 158.2
Quantity 25 mg
Formula Weight 158.0732 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.