missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-nitroaniline, 97%

Product Code. 10239823 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10239823 100 g Glass bottle 100g
10153630 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10239823 Supplier Thermo Scientific Acros Supplier No. 152021000
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 603-83-8 | C7H8N2O2 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-83-8
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007731
InChI Key HFCFJYRLBAANKN-UHFFFAOYSA-N
Synonym 2-amino-6-nitrotoluene, 3-nitro-o-toluidine, benzenamine, 2-methyl-3-nitro, o-toluidine, 3-nitro, 2-methyl-3-nitro-aniline, 1-amino-2-methyl-3-nitrobenzene, 2-nitro-6-aminotoluene, 2-methyl-3-nitrobenzenamine, 2-methyl-3-nitro-phenylamine, 2-methyl-3-nitrophenyl amine
PubChem CID 11783
IUPAC Name 2-methyl-3-nitroaniline
SMILES CC1=C(N)C=CC=C1[N+]([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 87°C to 91°C
Color Green-Yellow to Yellow
Boiling Point 305°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C6H3(NO2)NH2
Quantity 100 g
Beilstein 12, 848
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in ethanol and ether
Formula Weight 152.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-3-nitroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.