missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitrobenzotrifluoride, 98%

Product Code. 15442349
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15442349 5 g 5g
15432349 1 g 1g
15452349 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15442349 Supplier Thermo Scientific Alfa Aesar Supplier No. H64924.06
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 89976-12-5 | C8H6F3NO2 | 205.136 g/mol

Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89976-12-5
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.136
MDL Number MFCD01631684
InChI Key SVQCVQCIZWSPPX-UHFFFAOYSA-N
Synonym 2-methyl-5-nitrobenzotrifluoride, 1-methyl-4-nitro-2-trifluoromethyl benzene, 2-trifluoromethyl-4-nitrotoluene, 4-nitro-2-trifluoromethyl toluene, 1-methyl-4-nitro-2-trifluoromethyl-benzene, benzene, 1-methyl-4-nitro-2-trifluoromethyl, 3-trifluoromethyl-4-methylnitrobenzene, 2-trifluoromethyl-1-methyl-4-nitrobenzene, pubchem4444
PubChem CID 2775447
IUPAC Name 1-methyl-4-nitro-2-(trifluoromethyl)benzene
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])C(F)(F)F

Specifications

Color White
Quantity 5 g
UN Number UN2811
Beilstein 2506013
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 205.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Methyl-5-nitrobenzotrifluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.