missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzothiazol-6-ol, 96%

Product Code. 15476677
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476677 1 g 1g
15496677 250 mg 250mg
15486677 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15476677 Supplier Thermo Scientific Alfa Aesar Supplier No. H33812.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68867-18-5 | C8H7NOS | 165.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68867-18-5
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00227234
InChI Key ROFBPPIQUBJMRO-UHFFFAOYSA-N
Synonym 2-methyl-6-benzothiazolol, 2-methylbenzo d thiazol-6-ol, 6-benzothiazolol, 2-methyl, 2-methylbenzothiazol-6-ol, 6-hydroxy-2-methylbenzothiazole, 6-hydroxy-2-methylbenzthiazole, pubchem17329, 2-methyl-benzothiazol-6-ol, 2-methyl-6-hydroxybenzothiazole, 6-hydroxy-2-methyl-benzothiazole
PubChem CID 759304
IUPAC Name 2-methyl-1,3-benzothiazol-6-ol
SMILES CC1=NC2=CC=C(O)C=C2S1

Specifications

Melting Point 159°C to 161°C
Quantity 1 g
Formula Weight 165.21
Percent Purity 96%
Chemical Name or Material 2-Methylbenzothiazol-6-ol
compliance-icons
Product Identifier
  • 2-Methylbenzothiazol-6-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 6-Hydroxy-2-methylbenzothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.