missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthol, 98%

Product Code. 13438388 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13438388 5 kg Plastic drum 5kg
10338603 250 g Plastic bottle 250g
10397470 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13438388 Supplier Thermo Scientific Acros Supplier No. 156970050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 135-19-3 | C10H8O | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-19-3
Molecular Formula C10H8O
Molecular Weight (g/mol) 144.17
MDL Number MFCD00004067
InChI Key JWAZRIHNYRIHIV-UHFFFAOYSA-N
Synonym 2-naphthol, 2-naphthalenol, beta-naphthol, 2-hydroxynaphthalene, betanaphthol, isonaphthol, developer bn, naphthol b, azogen developer a, beta-napthol
PubChem CID 8663
ChEBI CHEBI:10432
IUPAC Name naphthalen-2-ol
SMILES C1=CC=C2C=C(C=CC2=C1)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 120.0°C to 124.0°C
Color White to Yellow
Boiling Point 285.0°C to 286.0°C
Flash Point 153°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic drum
Linear Formula C10H7OH
Quantity 5 kg
Beilstein 06,627
Merck Index 15,6469
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: 1 g/80mL boiling water, 1 g/0.8mL alcohol, 1 g/17mL chloroform, 1 g/1.3mL ether, soluble in ethanol, ether, chloroform, glycerol
Formula Weight 144.17
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Naphthol
Show More Show Less
compliance-icons compliance-icons
Identifiant produit
  • 2-Naphthol
Mention d'avertissement
  • Warning
Catégorie de danger
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
Mention de danger
  • H400-Very toxic to aquatic life.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Conseils de prudence
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.