missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthol, TRC

Product Code. 30004827
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004827 25 g 25g
30009299 5 g 5g
30011262 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004827 Supplier Toronto Research Chemicals Supplier No. TRCN36801525G
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-19-3
Molecular Formula C10 H8 O
Molecular Weight (g/mol) 144.17
Synonym Naphthalen-2-ol, Tolnaftate Imp. A (EP), beta-Naphthol, 2-Naphthalenol, 2-Naphthol (8CI), 2-Hydroxynaphthalene, Azogen Developer A, Betanaphthol, Bordeaux Base GP, C.I. 37500, C.I. Azoic Coupling Component 1, C.I. Developer 5, Developer A, Developer AMS, Developer BN, Developer NA, Isonaphthol, NSC 2044, NSC 5737, Naphthol B, β-Hydroxynaphthalene, β-Naphthol, β-Naphthyl alcohol
IUPAC Name naphthalen-2-ol
SMILES Oc1ccc2ccccc2c1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H8O/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7,11H
Formula Weight 144.0575
Chemical Name or Material beta-Naphthol
Product Title
Select an issue

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.