missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenyl phenyl ether, 99%

Product Code. 11409236
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11409236

Brand: Thermo Scientific Alfa Aesar A18088.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2216-12-8 | C12H9NO3 | 215.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2216-12-8
MDL Number MFCD00035744

Specifications

Density 1.23
Boiling Point 184°C (8 mmHg)
Flash Point 181°C (358°F)
Molecular Formula C10H18O
Refractive Index 1.6 00
Quantity 10 g
Beilstein 2051914
Synonym 2-Nitrodiphenyl ether; 2-Phenoxynitrobenzene
InChI Key IAIHUHQCLTYTSF-MRTMQBJTSA-N
SMILES CC1(C2CCC(C2)(C1O)C)C
IUPAC Name (1S,3R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-ol
Molecular Weight (g/mol) 154.25
PubChem CID 6997371
Formula Weight 215.21
Percent Purity 99%
Chemical Name or Material 2-Nitrophenyl phenyl ether
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.