missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroterephthalic acid 4-methyl ester, 97%

Product Code. 11372959
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372959 5 g 5g
11382959 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372959 Supplier Thermo Scientific Alfa Aesar Supplier No. H27779.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55737-66-1 | C9H7NO6 | 225.156 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55737-66-1
Molecular Formula C9H7NO6
Molecular Weight (g/mol) 225.156
MDL Number MFCD06203344
InChI Key VULISSQANNKDCH-UHFFFAOYSA-N
Synonym 4-methoxycarbonyl-2-nitrobenzoic acid, 2-nitro-4-methoxycarbonyl benzoic acid, 2-nitroterephthalic acid 4-methyl ester, acmc-1awlg, methyl 4-carboxy-3-nitrobenzoate, 4-carbomethoxy-2-nitrobenzoic acid, 4-methoxycarbonyl-2-nitrobenzoicacid, 2-nitro-terephthalic acid 4-methyl ester
PubChem CID 21906474
SMILES COC(=O)C1=CC(=C(C=C1)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 132°C to 136°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 225.17
Percent Purity 97%
Chemical Name or Material 2-Nitroterephthalic acid 4-methyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.