missing translation for 'onlineSavingsMsg'
Learn More

2-Oxo-4-phenylbutanoic Acid, Mikromol™

Product Code. 30051546
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051546 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051546 Supplier Mikromol Supplier No. MM0598.090025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Oxo-4-phenylbutanoic Acid
TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material 2-Oxo-4-phenylbutanoic Acid
CAS 710-11-2
Analyte or Component Names 2-Oxo-4-phenylbutanoic Acid
Shipping Condition Ice pack (-20°C)
Molecular Formula C10 H10 O3
Certifications/Compliance ISO 9001
Synonym Benzenebutanoic acid, α-oxo-, Butyric acid, 2-oxo-4-phenyl- (6CI,7CI,8CI), α-Oxobenzenebutanoic acid, 2-Oxo-4-phenylbutyric acid, 4-Phenyl-2-oxobutanoic acid, 4-Phenyl-2-oxobutyric acid, Benzylpyruvic acid
InChI Formula InChI=1S/C10H10O3/c11-9(10(12)13)7-6-8-4-2-1-3-5-8/h1-5H,6-7H2,(H,12,13)
SMILES OC(=O)C(=O)CCc1ccccc1
IUPAC Name 2-oxo-4-phenylbutanoic acid
Molecular Weight (g/mol) 178.1846
Quantity 25 mg
Formula Weight 178.063 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.