missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)aniline, Mikromol™

Product Code. 30051320
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051320 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051320 Supplier Mikromol Supplier No. MM1382.120100
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-(Trifluoromethoxy)aniline
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Trifluoromethoxy)aniline
CAS 1535-75-7
Analyte or Component Names 2-(Trifluoromethoxy)aniline
Shipping Condition Room Temperature
Molecular Formula C7 H6 F3 N O
Certifications/Compliance ISO 17025
Synonym Benzenamine, 2-(trifluoromethoxy)-, o-Anisidine, α,α,α-trifluoro- (6CI,7CI,8CI), 2-(Trifluoromethoxy)benzenamine, 2-(Trifluoromethyloxy)aniline, 2-Trifluoromethoxyaniline, 2-Trifluoromethoxyphenylamine, o-(Trifluoromethoxy)aniline, o-(Trifluoromethoxy)phenylamine
InChI Formula InChI=1S/C7H6F3NO/c8-7(9,10)12-6-4-2-1-3-5(6)11/h1-4H,11H2
SMILES Nc1ccccc1OC(F)(F)F
IUPAC Name 2-(trifluoromethoxy)aniline
Molecular Weight (g/mol) 177.12
Quantity 100 mg
Formula Weight 177.0401 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.